Learn More
Description
- Stands up to the toughest applications
- Designed for areas with high foot and forklift traffic
- Universal absorbents are strengthened by a tear-resistant barrier backing
Specifications
Specifications
| Absorbancy | 10.1 gl/gs |
| Color | Gray |
| Capacity (English) | 18.3 gal. |
| Material | Polypropylene |
| For Use With (Application) | High traffic areas where the risk for slips, trips and falls is greatest |
| Packaging Quantity | 100 Pads/Pk. |
| Dimensions (L x W) | 19 x 15 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.