Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ High Visibility Thermofill™ Lined Drivers Gloves

Catalog No. 17200173 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-173 Large
17-200-171 Small
17-200-172 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-200-173 Supplier Wells Lamont™ Supplier No. VPY0026L
  • $163.00
    $111.65 / Pack of 6

    Save $51.35

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Driver gloves are made from pigskin and provide comfort, warmth, and protection in colder environments.

  • Premium pigskin leather palm with orange fabric back.
  • Silver reflective knuckle strap.
  • Thermofill lined for colder environments.
  • Knit wrist construction prevents dirt and debris from entering the glove.
  • Sold by the dozen pair.

Specifications

Chemotherapeutic Rated No
Chlorinated No
Construction Cut and Sewn
Description Silver Reflective Knuckle Strap Adds Visibility
Color Hi-vis Orange
Grade Standard
Hand Specific
Material Top Grain Pigskin Leather
Product Type Leather Palm Gloves
Size Large
Style Gunn
For Use With (Application) Warehousing, Construction, Freezers, Outdoors
Gender Unisex
Glove Cuff Style Long Knitwrist
Grip Design Smooth
Hem Color Beige
Series VPY0026
Thumb Style Wing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.