Learn More
Lakeland Industries High Visibility Vests
Supplier:  Lakeland Industries V8A0112VLXL
Encompass_Preferred
Class 2, FR/AR Modacrylic Solid Hi-Vis Vest Hook and loop closure. Available in Hi-Vis Yellow and Hi-Vis Orange Built with GlenGuard? Hi-Vis fabric which is patented and inherently flame resistant. Designed to make safety vests cooler and more comfortable without sacrificing protection. The 2" 3M Scotchlite Retroreflectivity Material helps the eye perceive objects in low-light conditions when illuminated by a light source. Hi-Vis Yellow meets ANSI 107-2015 Class 2 Hi-Vis Orange meets ANSI 107-2010 Class 2 2" 3M Scotchlite Reflective Material Hook and loop closure 27" length
Catalog No. 50-284-1136