Learn More
MOLECULAR DEPOT LLC High Vitamin B12 - 50 mL
Catalog No. 5000013378
Encompass_Preferred
High Vitamin B12 Buffered Solution is a liquid-stable buffered solution containing high levels of vitamin B12 (2000-3000 pg/mL) This product has been used in the research and development of Vitamin B12 high and low working solutions This product is for in vitro research use only
Catalog No. 50-000-13378 Supplier MOLECULAR DEPOT LLC Supplier No. B2010582
High Vitamin B12 Buffered Solution is a liquid-stable buffered solution containing high levels of vitamin B12 (2000-3000 pg/mL) This product has been used in the research and development of Vitamin B12 high and low working solutions This product is for in vitro research use only