Learn More
MOLECULAR DEPOT LLC High Vitamin D - 50 mL
Catalog No. 5000013561
Encompass_Preferred
High Vitamin D Serum (50 mL) is human serum containing high levels of 25-OH vitamin D (200 ng/mL of total 25-OH vitamin D) This product is useful for the dilution of 25-OH vitamin D solutions as well as the research and development of 25-OH vitamin D high and low working solutions This product is liquid stable For in vitro research use only
Catalog No. 50-000-13561 Supplier MOLECULAR DEPOT LLC Supplier No. S2010004
High Vitamin D Serum (50 mL) is human serum containing high levels of 25-OH vitamin D (200 ng/mL of total 25-OH vitamin D) This product is useful for the dilution of 25-OH vitamin D solutions as well as the research and development of 25-OH vitamin D high and low working solutions This product is liquid stable For in vitro research use only