Learn More
OccuNomix™ High-Viz Rain Pants
Catalog No. 19520349 Shop All Occunomix International Inc Products
Click to view available options
Size:
Medium
Large
4X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-520-349 5X-Large
19-520-348 4X-Large
19-520-342 -
19-520-344 Large
19-520-343 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-520-349 Supplier OccuNomix™ Supplier No. LUXTENRY5X
This item is not returnable. View return policy
Lightweight and breathable for ease in mobility
Made with waterproof, windproof fabric. Feature 2 in. (5.08cm) 3M™ Scotchlite™ silver reflective material.
Pants
  • Lightweight ANSI Class E materials
  • Two pass-through pockets
  • Black fabric at ankles hides dirt and mud
Compliance
Meets ANSI/ISEA 107-2010 standards.
Specifications
Size 5X-Large
Product Type Hi-vis Rainwear Pants
Material Polyester, Polyurethane
Color Yellow
No. of Pockets 2
Certifications/Compliance ANSI Class E
Size (English) Waist 60 to 62 in.
Size (Metric) Waist 152.4 to 157.5 cm