Learn More
Euthanex High Volume Induction Chamber for use in Fume Hood, 11.5"" L x 7.5"" W x 8.0"" H.
Catalog No. 501540168
Encompass_Preferred
High Volume Induction Chamber. These chambers are typically for use under fume hoods or biosafety cabinets. They are not hinged like other chambers. The double lid design lifts off the chamber by pulling straight up. Chamber accommodates up to 16 mice or 4 medium rats. Constructed of heavy-duty polycarbonate plastic. Size: 11.5" L x 7.5" W x 8.0" H.
Catalog No. 50-154-0168 Supplier Euthanex Supplier No. EZ161A
High Volume Induction Chamber. These chambers are typically for use under fume hoods or biosafety cabinets. They are not hinged like other chambers. The double lid design lifts off the chamber by pulling straight up. Chamber accommodates up to 16 mice or 4 medium rats. Constructed of heavy-duty polycarbonate plastic. Size: 11.5" L x 7.5" W x 8.0" H.