Learn More
World Precision Instrument High Volume, Non-Gasket Chamber
SDP
Catalog No. 502538843
Encompass
  • For use with EZ-Anesthesia
  • For use with a hood or bio safety cabinet
  • Holds up to 16 mice or 4 med. rats
  • Heavy-duty, polycarbonate plastic
Catalog No. 50-253-8843 Supplier World Precision Instrument Supplier No. EZ161A
  • For use with EZ-Anesthesia
  • For use with a hood or bio safety cabinet
  • Holds up to 16 mice or 4 med. rats
  • Heavy-duty, polycarbonate plastic