Learn More
TA Instruments High Volume Sample Pan Die Set, for Standard DSC Press, Used to Seal High Volume Pans, Lids, and Seals (P/N 900825.902), Compatible with High Volume Sample Pan Kit
Catalog No. 5000580885 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Volume Sample Pan Die Set is designed for use with the Standard DSC Press to seal high-volume sample pans, lids, and seals (P/N: 900825.902). This die set ensures secure and consistent sealing for thermal analysis of aqueous-based or reactive materials. It is an essential component for preparing samples using the High Volume Sample Pan Kit and supports reliable performance in Waters TA Instruments DSC systems.
Catalog No. 50-005-80885 Supplier TA Instruments Supplier No. 900824.901
Waters TA Instruments High Volume Sample Pan Die Set is designed for use with the Standard DSC Press to seal high-volume sample pans, lids, and seals (P/N: 900825.902). This die set ensures secure and consistent sealing for thermal analysis of aqueous-based or reactive materials. It is an essential component for preparing samples using the High Volume Sample Pan Kit and supports reliable performance in Waters TA Instruments DSC systems.