Learn More
TA Instruments High Volume Sample Pan Die Set, Use with Standard DSC Press, Seal High Volume Sample Pans Lids and Seals, Accurate DSC Measurements, Standard DSC Press Die Set
Catalog No. 5000580863 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Volume Sample Pan Die Set for the Standard DSC Press is used to seal high-volume sample pans, lids, and seals in the Standard DSC Press for accurate DSC measurements.
Catalog No. 50-005-80863 Supplier TA Instruments Supplier No. 900790.901
Waters TA Instruments High Volume Sample Pan Die Set for the Standard DSC Press is used to seal high-volume sample pans, lids, and seals in the Standard DSC Press for accurate DSC measurements.