Learn More
TA Instruments High Volume Sample Pan Kit, 100 High Volume DSC Pans 100µL, Lids and O Ring Seals, Die Set for DSC Sample Press, Stainless Steel Needlepoint Tweezers, Instructions Included, Aqueous Based Pharmaceutical and Biological Materials
Catalog No. 5000580877 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Volume Sample Pan Kit contains 100 high-volume DSC pans (100 µL), lids, and o-ring seals, die set for DSC sample press, stainless steel needlepoint tweezers, and instructions. Ideal for aqueous-based pharmaceutical or biological materials.
Catalog No. 50-005-80877 Supplier TA Instruments Supplier No. 900860.901
Waters TA Instruments High Volume Sample Pan Kit contains 100 high-volume DSC pans (100 µL), lids, and o-ring seals, die set for DSC sample press, stainless steel needlepoint tweezers, and instructions. Ideal for aqueous-based pharmaceutical or biological materials.