Learn More
TA Instruments High Volume Sample Pan Kit, Includes 100 Pans (100µL), Lids, O-Ring Seals, Die Set, Tweezers, Instructions, Ideal for Aqueous-Based Materials and Condensation Reactions, Compatible with DSC Sample Press
Catalog No. 5000580876 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Volume Sample Pan Kit includes 100 high-volume DSC pans (100 µL), lids, o-ring seals, a die set for sealing pans using the DSC sample press, stainless steel needlepoint tweezers, and instructions. These pans are ideal for thermal analysis of pharmaceutical or biological materials that are aqueous-based. They are also excellent for measuring the heat of reaction in condensation-type curing processes. Designed for compatibility with Waters TA Instruments DSC systems and sample press tools.
Catalog No. 50-005-80876 Supplier TA Instruments Supplier No. 900825.901
Waters TA Instruments High Volume Sample Pan Kit includes 100 high-volume DSC pans (100 µL), lids, o-ring seals, a die set for sealing pans using the DSC sample press, stainless steel needlepoint tweezers, and instructions. These pans are ideal for thermal analysis of pharmaceutical or biological materials that are aqueous-based. They are also excellent for measuring the heat of reaction in condensation-type curing processes. Designed for compatibility with Waters TA Instruments DSC systems and sample press tools.