Learn More
TA Instruments High Volume Sample Pans, Lids, and Seals, Package of 100, Suppress Solvent Vaporization to 250°C, Requires High Volume Sample Pan Kit (P/N 900825.901), Compatible with DSC Sample Press
Catalog No. 5000580810 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Volume Sample Pans, Lids, and Seals (pkg. of 100) includes 100 sets of high-volume DSC pans (100 µL), lids, and seals designed to suppress solvent vaporization up to 250 °C. These components are ideal for aqueous-based sample analysis and require the High Volume Sample Pan Kit (P/N: 900825.901) for proper sealing and operation. Compatible with Waters TA Instruments DSC sample press, they support accurate and repeatable thermal measurements.
Catalog No. 50-005-80810 Supplier TA Instruments Supplier No. 900825.902
Waters TA Instruments High Volume Sample Pans, Lids, and Seals (pkg. of 100) includes 100 sets of high-volume DSC pans (100 µL), lids, and seals designed to suppress solvent vaporization up to 250 °C. These components are ideal for aqueous-based sample analysis and require the High Volume Sample Pan Kit (P/N: 900825.901) for proper sealing and operation. Compatible with Waters TA Instruments DSC sample press, they support accurate and repeatable thermal measurements.