Learn More
Cayman Chemical HIghBIndng 96 SolId Pl 5ea
SDP
Catalog No. 502942121
Encompass_Preferred
A fluorescent dGTP derivative selectively inhibits recombinant mammalian AC1 AC2 AC5 and AC6 (Kis 260 1200 420 and 470 nM respectively) bovine lung sGC (Ki 220 nM) and B. anthracis edema factor AC toxin (Ki 630 nM) over rat testis sAC and B. pertussis AC toxin (Kis 43 and 14 UM respectively) has been used to study the activity of various GTPases including EF-Tu and H-Ras p21
Catalog No. 50-294-2121 Supplier Cayman Chemical Supplier No. 4000105 EA
A fluorescent dGTP derivative selectively inhibits recombinant mammalian AC1 AC2 AC5 and AC6 (Kis 260 1200 420 and 470 nM respectively) bovine lung sGC (Ki 220 nM) and B. anthracis edema factor AC toxin (Ki 630 nM) over rat testis sAC and B. pertussis AC toxin (Kis 43 and 14 UM respectively) has been used to study the activity of various GTPases including EF-Tu and H-Ras p21