Learn More
Enzo Life Sciences HIGHDEF black IHC chromogen (HRP) (30 ml)
SDP
Catalog No. NC1609441
Encompass_Preferred
Unique high definition black IHC chromogen for HRP-activated development. HIGHDEF black IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridization systems. With at least 1 day working stability, this substrate/chromogen system can be used in any automation system as well as manual use. Contents: HIGHDEF black IHC chromogen (HRP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC1609441 Supplier Enzo Life Sciences Supplier No. ADI9501710030
Unique high definition black IHC chromogen for HRP-activated development. HIGHDEF black IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridization systems. With at least 1 day working stability, this substrate/chromogen system can be used in any automation system as well as manual use. Contents: HIGHDEF black IHC chromogen (HRP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C