Learn More
Enzo Life Sciences HIGHDEF blue IHC chromogen (AP) (30 ml)
SDP
Catalog No. NC1609439
Encompass_Preferred
High definition IHC chromogen producing brilliant blue color for AP-activated development. HIGHDEF blue IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems. Contents: HIGHDEF blue IHC chromogen (AP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC1609439 Supplier Enzo Life Sciences Supplier No. ADI9501500030
High definition IHC chromogen producing brilliant blue color for AP-activated development. HIGHDEF blue IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems. Contents: HIGHDEF blue IHC chromogen (AP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C