Learn More
Enzo Life Sciences HIGHDEF Green AP Chromogen/Substrate (30 ml)
SDP
Catalog No. NC1609432
Encompass_Preferred
High definition IHC chromogen producing brilliant green color for AP-activated development. HIGHDEF Green AP Chromogen/Substrate is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems. Contents: HIGHDEF Green AP Chromogen, HIGHDEF Green AP Substrate. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC1609432 Supplier Enzo Life Sciences Supplier No. ENZACC1300030
High definition IHC chromogen producing brilliant green color for AP-activated development. HIGHDEF Green AP Chromogen/Substrate is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems. Contents: HIGHDEF Green AP Chromogen, HIGHDEF Green AP Substrate. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C