Learn More
Enzo Life Sciences HIGHDEF Hematoxylin (100ml)
SDP
Catalog No. 502013355
Encompass_Preferred
HIGHDEF Hematoxylin is intended for use in the histologic demonstration of nuclear staining. This staining technique is sometimes used to make the critical distinction between a normal nucleus and an abnormal one. Formulation: Enhancers added to improve signal. Long Term Storage: Ambient. Applications: IHC ISH (in situ hybridization).
Catalog No. 50-201-3355 Supplier Enzo Life Sciences Supplier No. ENZACC1060100
HIGHDEF Hematoxylin is intended for use in the histologic demonstration of nuclear staining. This staining technique is sometimes used to make the critical distinction between a normal nucleus and an abnormal one. Formulation: Enhancers added to improve signal. Long Term Storage: Ambient. Applications: IHC ISH (in situ hybridization).