Learn More
Enzo Life Sciences HIGHDEF IHC fluoromount (25ml)
SDP
Catalog No. 502007121
Encompass_Preferred
Permanent, high stability mounting medium for high definition visualization of IHC. HIGHDEF IHC fluoromount is for mounting tissue specimens stained with fluorescent dyes. Designed for use in fluorescence, but can be used with other applications. Formulation: Liquid. Mounting medium containing <0.1% sodium azide. Long Term Storage: Ambient. Applications: IF, IHC ISH (in situ hybridization).
Catalog No. 50-200-7121 Supplier Enzo Life Sciences Supplier No. ADI9502600025
Permanent, high stability mounting medium for high definition visualization of IHC. HIGHDEF IHC fluoromount is for mounting tissue specimens stained with fluorescent dyes. Designed for use in fluorescence, but can be used with other applications. Formulation: Liquid. Mounting medium containing <0.1% sodium azide. Long Term Storage: Ambient. Applications: IF, IHC ISH (in situ hybridization).