Learn More
Enzo Life Sciences HIGHDEF IHC mount
SDP
Catalog No. NC1692622
Encompass_Preferred
HIGHDEF IHC mount is an aqueous-based permanent mounting medium designed for use with chromogenic labeling. Use for mounting of tissue specimens stained with peroxidase and alkaline phosphatase-based systems and various fluorescent dyes.
Catalog No. NC1692622 Supplier Enzo Life Sciences Supplier No. ADI9502610030
HIGHDEF IHC mount is an aqueous-based permanent mounting medium designed for use with chromogenic labeling. Use for mounting of tissue specimens stained with peroxidase and alkaline phosphatase-based systems and various fluorescent dyes.