Learn More
Enzo Life Sciences HIGHDEF red IHC chromogen (AP, plus) (30 ml)
SDP
Catalog No. NC1392004
Encompass_Preferred
High definition IHC chromogen producing brilliant red color for AP-activated development in automated and manual systems. HIGHDEF red IHC chromogen (AP, plus) is intended to be used in conjuction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems that run on automated staining instruments as well as manual use. Contents: HIGHDEF red IHC chromogen (AP, plus) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC1392004 Supplier Enzo Life Sciences Supplier No. ADI9501410030
High definition IHC chromogen producing brilliant red color for AP-activated development in automated and manual systems. HIGHDEF red IHC chromogen (AP, plus) is intended to be used in conjuction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems that run on automated staining instruments as well as manual use. Contents: HIGHDEF red IHC chromogen (AP, plus) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C