Learn More
Enzo Life Sciences HIGHDEF Red IHC Chromogen (HRP), (30ml)
SDP
Catalog No. NC1609433
Encompass_Preferred
The HIGHDEF Red IHC Chromogen can be used in conjunction with peroxidase-based immunostaining systems. Peroxidase from the immunodetection system reacts with hydrogen peroxide substrate to degrade it, which reacts with AEC, precipitating it at positive sites yielding a reddish brown colored product. Appearance: Amber solution. Long Term Storage: +4°C
Catalog No. NC1609433 Supplier Enzo Life Sciences Supplier No. ADI9502100030
The HIGHDEF Red IHC Chromogen can be used in conjunction with peroxidase-based immunostaining systems. Peroxidase from the immunodetection system reacts with hydrogen peroxide substrate to degrade it, which reacts with AEC, precipitating it at positive sites yielding a reddish brown colored product. Appearance: Amber solution. Long Term Storage: +4°C