Learn More
Research Products International Corp High Density Polyethylene Vials, 20ml Capacity, 22mm Poly Lined Plastic Screw Caps, White, 500 per Case
SDP
Catalog No. 50212998
Encompass
Economical and efficient alternative to glass vials. Major advantages include higher efficiency, lower background and overall higher performance. Manufactured from an improved high density polyethylene blend of polyethylene that resists solvent permeation. Biodegradable solvent based cocktails can be stored indefinitely in these vials.
Catalog No. 50-212-998 Supplier Research Products International Corp Supplier No. 121043
Economical and efficient alternative to glass vials. Major advantages include higher efficiency, lower background and overall higher performance. Manufactured from an improved high density polyethylene blend of polyethylene that resists solvent permeation. Biodegradable solvent based cocktails can be stored indefinitely in these vials.