Learn More
Ted Pella Inc Highest Grade V1 AFM Mica Discs, 12mm, 10/pk
SDP
Catalog No. NC9971063
Encompass
Highest quality grade V1 mica, 0.21mm (0.0085”) thick. Interleaved, in packages of 10. Can be mounted on AFM/STM discs.
This item is not returnable. View return policy
Catalog No. NC9971063 Supplier Ted Pella Inc Supplier No. 5012
Highest quality grade V1 mica, 0.21mm (0.0085”) thick. Interleaved, in packages of 10. Can be mounted on AFM/STM discs.