Learn More
Qosina Corp HIGHFLOW CHECK VLVE,BARBED,1/4
Catalog No. NC2245842
Encompass
High-Flow Check Valve, BarbedFits 1/4 inch ID Tubing (0.25 inch, 6.4 mm); 0.075 psi CP
This item is not returnable. View return policy
Catalog No. NC2245842 Supplier Qosina Corp Supplier No. 91050
High-Flow Check Valve, BarbedFits 1/4 inch ID Tubing (0.25 inch, 6.4 mm); 0.075 psi CP