Learn More
Adam Equipment Adam Equipment Highland Approved Portable Precision Balance HCB 103aM 600g capacity x d=0.001g e=0.01g readability
Supplier:  Adam Equipment HCB 103AM
Encompass_Preferred
Adam Equipment Highland Approved Portable Precision Balances are packed with features and meet Class II requirements for use in retail applications. Readings are provided in carats, grams, or kilograms. The Highland includes RS-232 and USB interfaces, rechargeable battery and AC operation, capacity tracker, adjustable feet and level, lockdown capability and more. The Highland's innovative design features a removable draft shield to minimize the effects of drafts. ShockProtect® helps prevent overloads to maintain smooth, uncompromised performance. With built-in applications for parts counting, percentage weighing and totalization / accumulation, the NTEP-approved Highland is an outstanding value for legal-for-trade applications. Highland approved portable precision balances contain practical features that simplify legal-for-trade measurement, lab work, field testing, and commercial use. AC adapter included. Rechargeable battery included
Catalog No. NC2612381