Learn More
Pyramex™ Highlander™ Safety Eyewear
Catalog No. 19155889 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Black, Green
Black, Red
Black, Silver
Lens Tint:
Blue, Mirror
Clear
Gray
Indoor/Outdoor, Mirror
Red, Lime
Silver, Mirror
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-889 Black Indoor/Outdoor, Mirror
19-155-885 Black Clear
19-155-886 Black Gray
19-155-887 Black Red, Lime
19-155-888 Black Silver, Mirror
19-155-909 Black, Green Clear
19-155-911 Black, Red Clear
19-155-912 Black, Red Gray
19-155-997 Black, Silver Gray
19-155-998 Black, Silver Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-155-889 Supplier Pyramex™ Supplier No. SBB5080DT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Soft foam carriage, durable frame with detachable strap
  • Durable frame with detachable strap
  • Inner lens clip provides soft foam for comfort fit and extreme styling
  • Can be worn with inner foam for additional dust protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Indoor/Outdoor, Mirror
    Frame Color Black
    Weight (Metric) 35 g
    Lens Material Polycarbonate
    Frame Material Nylon