Learn More
Thermo Scientific™ HIGHPlate 6000 Microplate Rotor for 4 L General Purpose X Pro Centrifuges
GSA_VA
Catalog No. 75003606 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
2 x 5 Standard Plates
Catalog No. Capacity
75-003-606 2 x 5 Standard Plates
1 options
Catalog No. 75-003-606 Supplier Thermo Scientific™ Supplier No. 75003606
  • $9,990.00
    $7,121.65 / Each of 1
    Save $2,868.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor.
Spin filter plates with the Thermo Scientific™ HIGHPlate™ 6000 Microplate Rotor, for use with DNA purification protocols.
Compatible with:
Thermo Scientific™ Sorvall™ Legend™ XT/XTR, Multifuge™ X3/X3R
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description HIGHPlate 6000 Microplate Rotor
Type Swinging Bucket Rotor
Capacity 2 x 5 Standard Plates
Max. RCF 6,168 x g
Max. Speed 6,300 rpm
Radius (Metric) 139 mm
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Angle 90°
Certifications/Compliance Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Biocontainment Yes