Learn More
Magbio Genomics Inc HighPrep DTR (5mL)
SDP
Catalog No. 501656586
Encompass_Preferred
HighPrep DTR is a high performance paramagnetic bead-based system for Big Dye sequencing clean up. Designed to remove unincorporated dye terminators from Sanger sequencing reaction, the HighPrep DTR system consists of a selective binding of DNA to the HighPrep DTR paramagnetic beads, followed with washing off nucleotides, primers and non-targeted amplicons, and finally elution of pure DNA. HighPrep DTR is designed for both manual and fully automated purification of sequencing products. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol.
Catalog No. 50-165-6586 Supplier Magbio Genomics Inc Supplier No. DT70005
HighPrep DTR is a high performance paramagnetic bead-based system for Big Dye sequencing clean up. Designed to remove unincorporated dye terminators from Sanger sequencing reaction, the HighPrep DTR system consists of a selective binding of DNA to the HighPrep DTR paramagnetic beads, followed with washing off nucleotides, primers and non-targeted amplicons, and finally elution of pure DNA. HighPrep DTR is designed for both manual and fully automated purification of sequencing products. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol.