Learn More
Magbio Genomics Inc HighPrep PCR (50mL)
SDP
Catalog No. 501656583
Encompass_Preferred
HighPrep™ PCR reagent is a post PCR and Next Gen library prep clean up system based on paramagnetic beads technology, designed for an efficient purification of PCR amplicons. The purification consists of removal of salts, primers, primer-dimers, dNTPs, as DNA fragments are selectively bound to the magnetic beads particles; and highly purified DNA is eluted with low salt elution buffer or water and can be used directly for downstream applications. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Caliper, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol as well as it can be performed manually.
Catalog No. 50-165-6583 Supplier Magbio Genomics Inc Supplier No. AC60050
HighPrep™ PCR reagent is a post PCR and Next Gen library prep clean up system based on paramagnetic beads technology, designed for an efficient purification of PCR amplicons. The purification consists of removal of salts, primers, primer-dimers, dNTPs, as DNA fragments are selectively bound to the magnetic beads particles; and highly purified DNA is eluted with low salt elution buffer or water and can be used directly for downstream applications. The protocol can be adapted to your current liquid handling workstation (e.g. Beckman, Hamilton, Tecan, Caliper, Perkin Elmer, Agilent and Eppendorf) utilizing your current protocol as well as it can be performed manually.