Learn More
Magbio Genomics Inc HighPrep RNA Elite (5mL)
SDP
Catalog No. 501656590
Encompass_Preferred
The HighPrep RNA Elite system utilizes MagBio’s solid-phase paramagnetic bead technology for high-throughput purifi cation of RNA or cDNA for in vitro applications such as transcription, antisense RNA (aRNA) amplifi cation as well as RNA and cDNA probe synthesis. This protocol enables recovery of micro RNA (miRNA), small RNA and total RNA from enzymatic reactions, concentrating miRNA and total RNA from a diluted sample. This protocol can be used for manual procedure as well as guideline for adapting the kit to automatic liquid handling instruments.
Catalog No. 50-165-6590 Supplier Magbio Genomics Inc Supplier No. RC90005
The HighPrep RNA Elite system utilizes MagBio’s solid-phase paramagnetic bead technology for high-throughput purifi cation of RNA or cDNA for in vitro applications such as transcription, antisense RNA (aRNA) amplifi cation as well as RNA and cDNA probe synthesis. This protocol enables recovery of micro RNA (miRNA), small RNA and total RNA from enzymatic reactions, concentrating miRNA and total RNA from a diluted sample. This protocol can be used for manual procedure as well as guideline for adapting the kit to automatic liquid handling instruments.