Learn More
McMaster-Carr High-Temperature Metallic Gasket
Catalog No. NC1458177
Encompass
High-Temperature Metallic Gasket for 2 Pipe Size, ANSI Class 300, Graphite Filler
Catalog No. NC1458177 Supplier McMaster-Carr Supplier No. 44955K176
High-Temperature Metallic Gasket for 2 Pipe Size, ANSI Class 300, Graphite Filler