Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Hill Manufacturing Hilco Lube, Multipurpose, Amber Aerosol, 12 Pack

Catalog No. 502870438
Encompass

No. 5035 Hilco Lube is a multipurpose aerosol lubricant designed for various applications. It is characterized by its amber appearance and petroleum odor, providing effective lubrication while being classified as non-flammable by the ASTM Flame Extension Test.

  • Contains gas under pressure, may explode if heated
  • Non-flammable according to ASTM Flame Extension Test
  • Initial boiling point and range 300°F
  • Flash point 105°F TCC
  • Specific gravity 0.8–0.9
  • Nil solubility in water
  • Amber aerosol with petroleum odor

Catalog No. 50-287-0438 Supplier MSC Supplier No. 80106610
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

No. 5035 Hilco Lube is a multipurpose aerosol lubricant designed for various applications. It is characterized by its amber appearance and petroleum odor, providing effective lubrication while being classified as non-flammable by the ASTM Flame Extension Test.

  • Contains gas under pressure, may explode if heated
  • Non-flammable according to ASTM Flame Extension Test
  • Initial boiling point and range 300°F
  • Flash point 105°F TCC
  • Specific gravity 0.8–0.9
  • Nil solubility in water
  • Amber aerosol with petroleum odor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.