Learn More
Cayman Chemical HIleProProOHtrIfluoroac 10mg
SDP
Catalog No. 502938768
Encompass_Preferred
A peptide ACE inhibitor (IC50 = 5 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, mRNA expression of IL-6 and IL-1β, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice
Catalog No. 50-293-8768 Supplier Cayman Chemical Supplier No. 2889910 MG
A peptide ACE inhibitor (IC50 = 5 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, mRNA expression of IL-6 and IL-1β, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice