Learn More
Cayman Chemical H-Ile-Pro-Pro-OH (trifluoroacetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502940364
Encompass_Preferred
A peptide ACE inhibitor (IC50 = 5 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, mRNA expression of IL-6 and IL-1β, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice
Catalog No. 50-294-0364 Supplier Cayman Chemical Supplier No. 2889950 MG
A peptide ACE inhibitor (IC50 = 5 µM); inhibits acetylcholine-induced NO production in HUVECs at 1 µM; induces vasorelaxation in precontracted isolated rat aortic rings; decreases systolic blood pressure in spontaneously hypertensive rats at 0.3 mg/kg; dietary administration reduces plasma levels of total cholesterol, HDL-cholesterol, and triglycerides, mRNA expression of IL-6 and IL-1β, and aortic arch intimal thickening and atherosclerotic plaque formation in ApoE-/- mice