Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation HILIC QC Reference Material, Liquid, 1 mL

Catalog No. 502037891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The HILIC QC Reference Material is a four-component mix designed as a standard to benchmark hydrophilic interaction liquid chromatography (HILIC) separations. It contains an un-retained void marker, a polar neutral compound, and two polar basic compounds, providing stable chromatographic separation for HILIC columns. This material assists in validating, monitoring, developing, and troubleshooting HILIC separations.

  • Designed for benchmarking HILIC separations.
  • Assesses and compares HILIC column performance.
  • Aids in validating, monitoring, and troubleshooting HILIC separations.
  • Supplied in a 1 mL vial for direct injection.
  • Compatible with various mobile phases and columns.
  • Contains Acenaphthene, Thymine, Adenine, and Cytosine.
  • Store at 4°C; 12 months shelf life.

Catalog No. 50-203-7891 Supplier Waters Corp Supplier No. 186007226
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The HILIC QC Reference Material is a four-component mix designed as a standard to benchmark hydrophilic interaction liquid chromatography (HILIC) separations. It contains an un-retained void marker, a polar neutral compound, and two polar basic compounds, providing stable chromatographic separation for HILIC columns. This material assists in validating, monitoring, developing, and troubleshooting HILIC separations.

  • Designed for benchmarking HILIC separations.
  • Assesses and compares HILIC column performance.
  • Aids in validating, monitoring, and troubleshooting HILIC separations.
  • Supplied in a 1 mL vial for direct injection.
  • Compatible with various mobile phases and columns.
  • Contains Acenaphthene, Thymine, Adenine, and Cytosine.
  • Store at 4°C; 12 months shelf life.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.