Learn More
Sigma Aldrich Fine Chemicals Biosciences HiLoadR 16/600 SuperdexR 2
Catalog No. 502908814
Encompass
HiLoad(R) 16/600 Superdex(R) 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.Superdex(R) prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges Mr < 10 000 (Superdex(R) 30 pg) Mr 3 000 to 70 000 (Superdex(R) 75 pg) and Mr 10 000 600 000 (Superdex(R) 200 pg). The medium combines high mechanical strength with high hydrophilicity allowing high flow rates and minimal nonspecific interactions.
Catalog No. 50-290-8814 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28989335
HiLoad(R) 16/600 Superdex(R) 200 prep grade are prepacked XK columns designed for high-resolution preparative gel filtration chromatography. The column is an excellent choice for separating MAbs from dimers and contaminants.Superdex(R) prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component. Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges Mr < 10 000 (Superdex(R) 30 pg) Mr 3 000 to 70 000 (Superdex(R) 75 pg) and Mr 10 000 600 000 (Superdex(R) 200 pg). The medium combines high mechanical strength with high hydrophilicity allowing high flow rates and minimal nonspecific interactions.