Learn More
Dojindo Molecular Technologies Inc HilyMax (1ML x 1) is a high transfection efficiency reagent, and is the best reagent for cell signaling research.
SDP
Catalog No. NC0645912
Encompass_Preferred
HilyMax is a gene transfection reagent that forms a liposome to be used for highly efficient gene transfection to a wide variety of cells. HilyMax readily interacts with DNA because cationic liposomes (+) and anionic DNA (-) spontaneously form DNA liposome complexes. The overall charge of DNA-HilyMax complex is positive, such that the DNA-HilyMax complex is electrostatically bound on an anionic cell surface and introduces DNA into the cell by endocytosis. Since the serum in the growth medium does not interfere with the transfection using HilyMax, no exchange of the medium during the transfection is required. HilyMax does not contain biological components that might interfere with the transfection.
Catalog No. NC0645912 Supplier Dojindo Molecular Technologies Inc Supplier No. H35710
HilyMax is a gene transfection reagent that forms a liposome to be used for highly efficient gene transfection to a wide variety of cells. HilyMax readily interacts with DNA because cationic liposomes (+) and anionic DNA (-) spontaneously form DNA liposome complexes. The overall charge of DNA-HilyMax complex is positive, such that the DNA-HilyMax complex is electrostatically bound on an anionic cell surface and introduces DNA into the cell by endocytosis. Since the serum in the growth medium does not interfere with the transfection using HilyMax, no exchange of the medium during the transfection is required. HilyMax does not contain biological components that might interfere with the transfection.