Learn More
AnaSpec Beta - Amyloid (1 - 42), HiLyteᵀᴹ Fluor 488 - labeled, Human. 0.1 mg
Supplier:  AnaSpec AS6047901
Encompass
This is a fluorescent (HiLyte™ Fluor 488)-labeled ß-Amyloid peptide, Abs/Em=503/528 nm. Hilyte 488™ Fluor labeled Aß (1-42) has a brighter intensity than FAM-labeled Aß (1-42). Purity % Peak Area By HPLC ≥ 95%; Molecular Weight 4870.5; Storage -20°C
Catalog No. 50-824-468