Learn More
Dojindo Molecular Technologies Inc HiLyte Fluor™ 555 Labeling Kit - NH2, 3 samples
SDP
Catalog No. NC3366712
Encompass
HiLyte Fluor* 555 Labeling Kit-NH2 is used mainly for the preparation of red fluorescence-labeled proteins, such as IgG, for immunostaining, and cellular proteins for tracing. NH2-reactive HiLyteFluor 555, a component of this kit, has a succinimidyl group (NHS) that reacts with amino groups on proteins or other molecules (Fig. 1). This kit contains all the reagents necessary for labeling. Each tube of HiLyte Fluor 555 can label up to 200 μg of IgG, conjugating about 4 to 6 HiLyte Fluor 555 molecules per IgG molecule. The labeling process is simple-add the NH2-reactive HiLyte Fluor 555 to IgG solution on a membrane and incubate at 37ºC for 10 minutes. The excess HiLyte Fluor 555 molecules can be removed by a filtration tube. The excitation and emission wavelengths of the HiLyte Fluor 555-labeled IgG are 555 nm and 570 nm, respectively (Fig. 2).* HiLyte Fluor is a trademark of AnaSpec, Inc.
Catalog No. NC3366712 Supplier Dojindo Molecular Technologies Inc Supplier No. LK1410
HiLyte Fluor* 555 Labeling Kit-NH2 is used mainly for the preparation of red fluorescence-labeled proteins, such as IgG, for immunostaining, and cellular proteins for tracing. NH2-reactive HiLyteFluor 555, a component of this kit, has a succinimidyl group (NHS) that reacts with amino groups on proteins or other molecules (Fig. 1). This kit contains all the reagents necessary for labeling. Each tube of HiLyte Fluor 555 can label up to 200 μg of IgG, conjugating about 4 to 6 HiLyte Fluor 555 molecules per IgG molecule. The labeling process is simple-add the NH2-reactive HiLyte Fluor 555 to IgG solution on a membrane and incubate at 37ºC for 10 minutes. The excess HiLyte Fluor 555 molecules can be removed by a filtration tube. The excitation and emission wavelengths of the HiLyte Fluor 555-labeled IgG are 555 nm and 570 nm, respectively (Fig. 2).* HiLyte Fluor is a trademark of AnaSpec, Inc.