Learn More
Enzo Life Sciences (+)-Himbacine (5mg). CAS: 6879-74-9
SDP
Supplier:  Enzo Life Sciences ALX550061M005
Encompass_Preferred
Potent muscarinic antagonist that displays selectivity for the M2 or M4 receptors. Alternative name: (3S,3aR,4R,4aS,8aR,9aS)-4-[(1E)-2-[(2R,6S)-1,6-Dimethyl-2-piperdinyl]ethenyl]decahydro-3-methyl-naphtho[2,3-c]furan-1(3H)-one. Purity: ≥98%. Appearance: White to off-white powder. Solubility: Soluble in methanol, 100% ethanol or methylene chloride; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-9471