Learn More
New England Biolabs, Inc. HincII – 5000 units
SDP
Catalog No. 50811924
Encompass
A restriction endonuclease that recognizes the sequence GTY_^RAC.
Catalog No. 50-811-924 Supplier New England Biolabs, Inc. Supplier No. R0103L
A restriction endonuclease that recognizes the sequence GTY_^RAC.