Learn More
Description
- Ready-to-use serum-free media used for the efficient differentiation of iPSC derived definitive endoderm (DE) cells to CDX-2+ hindgut endoderm cells
- 4 day differentiation protocol has been validated on two human iPS cell lines
- Differentiated cells express high levels of the hindgut endoderm marker, CDX-2
- CDX-2 hindgut endoderm cells can be differentiated into specialized lineages such as intestinal colon organoids after embedding in Matrigel and cultured in 3dGRO™ Human Colon Organoid Expansion Medium
- No growth
Specifications
Specifications
| Content And Storage | Upon receipt -20 °C, Ready to use 2 to 8 ° overnight thaw, Use immediately store (at 2°C to 8°C up to 1 week, Do not re-freeze) |
| Product Type | Induction Medium |
| Quantity | 50 mL |
| Solution Type | Buffer |
| Endotoxin Level | < EU/mL |
| For Use With (Application) | Stem Cell Culture |
| pH | 7.0 to 7.4 |
| Form | Liquid |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.