Learn More
New England Biolabs, Inc. HindIII – 10000 units
SDP
Catalog No. 50811929
Encompass
A restriction endonuclease that recognizes the sequence A^AGCT_T.
Catalog No. 50-811-929 Supplier New England Biolabs, Inc. Supplier No. R0104T
A restriction endonuclease that recognizes the sequence A^AGCT_T.