Learn More
New England Biolabs, Inc. HindIII – 50000 units
SDP
Catalog No. 50811927
Encompass
A restriction endonuclease that recognizes the sequence A^AGCT_T.
Catalog No. 50-811-927 Supplier New England Biolabs, Inc. Supplier No. R0104M
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence A^AGCT_T.