Learn More
New England Biolabs, Inc. HindIII – 50000 units
SDP
Catalog No. 50811926
Encompass
A restriction endonuclease that recognizes the sequence A^AGCT_T.
Catalog No. 50-811-926 Supplier New England Biolabs, Inc. Supplier No. R0104L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence A^AGCT_T.