Learn More
New England Biolabs, Inc. HinfI – 5000 units
SDP
Catalog No. 50812052
Encompass
A restriction endonuclease that recognizes the sequence G^ANT_C.
Catalog No. 50-812-052 Supplier New England Biolabs, Inc. Supplier No. R0155S
A restriction endonuclease that recognizes the sequence G^ANT_C.