Learn More
New England Biolabs, Inc. HinfI – 25000 units
SDP
Catalog No. 50812051
Encompass
A restriction endonuclease that recognizes the sequence G^ANT_C.
Catalog No. 50-812-051 Supplier New England Biolabs, Inc. Supplier No. R0155M
A restriction endonuclease that recognizes the sequence G^ANT_C.