Learn More
New England Biolabs, Inc. HinfI – 5000 units
SDP
Supplier:  New England Biolabs, Inc. R0155T
Encompass
A restriction endonuclease that recognizes the sequence G^ANT_C.
Catalog No. 50-812-053