Learn More
Ideal Vacuum Products LLC HINGE CLAMP KF-16 VACUUM FITTI
SDP
Catalog No. NC1992660
Encompass
Hinge Clamp KF-10, KF-16 Vacuum Fittings, ISO-KF Flange Size NW10, NW-16, Aluminum. These KF-10, KF-16 Hinge Clamp vacuum fittings follow ISO-KF standards and have a flange size of NW-10, NW-16. They are commonly used to build foreline vacuum plumbing and process systems. They are often called quick flange fittings because they are quick to assemble. A circumferential clamp and centering ring form the vacuum seal between the connecting flanges (please see top right figure - click to enlarge). The centering ring contains a rubber elastomer o-ring. The standard sizes for quick flange fittings are KF-10, KF-16, KF-25, KF-40, and KF-50 with flange sizes NW-10, NW-16, NW-25, NW-40, and NW-50, respectively. They are made of Aluminum.
Catalog No. NC1992660 Supplier Ideal Vacuum Products LLC Supplier No. P101198
Hinge Clamp KF-10, KF-16 Vacuum Fittings, ISO-KF Flange Size NW10, NW-16, Aluminum. These KF-10, KF-16 Hinge Clamp vacuum fittings follow ISO-KF standards and have a flange size of NW-10, NW-16. They are commonly used to build foreline vacuum plumbing and process systems. They are often called quick flange fittings because they are quick to assemble. A circumferential clamp and centering ring form the vacuum seal between the connecting flanges (please see top right figure - click to enlarge). The centering ring contains a rubber elastomer o-ring. The standard sizes for quick flange fittings are KF-10, KF-16, KF-25, KF-40, and KF-50 with flange sizes NW-10, NW-16, NW-25, NW-40, and NW-50, respectively. They are made of Aluminum.